D-α-Phenylalanin
Lieferant: BACHEM BIOCHEMICA
Synonyme:
D-Phenylalanin, (R)-(+)-2-Amino-3-phenylpropansäure, 3-Phenyl-D-alanin, H-D-Phe-OH, 3-Phenyl-D(+)-alanin, (R)-(+)-2-Amino-3-phenylpropionsäure, (R)-(+)-2-Amino-3-phenylpropionic acid
4031149.0025
4031149.0100
BACE4031149.0025EA
67.2
EUR
BACE4031149.0025
BACE4031149.0100
D-α-Phenylalanin
D-α-Phenylalanin
Formel:
C₉H₁₁NO₂ Molecular Weight: 165.19 g/mol Siedepunkt: 295 |
MDL:
MFCD00004270 CAS-Nummer: 673-06-3 EINECS: 211-603-5 Merck Index: 12,07425 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...