L-α-Phenylalanin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
H-Phe-OH, H-L-Phe-OH, L-α-Phenylalanin, 3-Phenyl-L(-)-alanin, 3-Phenyl-L-alanin, (S)-(-)-2-Amino-3-phenylpropansäure
78019-100G
78019-25G
78019-500G
SIAL78019-100GEA
197
EUR
SIAL78019-100G
SIAL78019-25G
SIAL78019-500G
L-α-Phenylalanin, Sigma-Aldrich®
L-α-Phenylalanin
Formel:
C₉H₁₁NO₂ Molecular Weight: 165,19 g/mol Siedepunkt: 313 °C (1013 hPa) Schmelzpunkt: 173 °C Dichte: 0,754 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064227 CAS-Nummer: 63-91-2 EINECS: 200-568-1 Merck Index: 14,07271 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...