4-Amino-3-brombenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

642770-1G 642770-5G
SIAL642770-1GEA 69 EUR
SIAL642770-1G SIAL642770-5G
4-Amino-3-brombenzonitril, Sigma-Aldrich®
4-Amino-3-brombenzonitril
Formel: C₇H₅BrN₂
Molecular Weight: 197,03 g/mol
MDL: MFCD01861386
CAS-Nummer: 50397-74-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR