2-Cyan-3-fluoranilin

Lieferant: Thermo Scientific

Synonyme: 2-Amino-6-fluorbenzonitril, 2-Amino-6-fluorbenzoesäurenitril

BTB07341.1 BTB07341.10 BTB07341.50
MAYBBTB07341.1EA 0 EUR
MAYBBTB07341.1 MAYBBTB07341.10 MAYBBTB07341.50
2-Cyan-3-fluoranilin
2-Cyan-3-fluoranilin
Formel: C₇H₅FN₂
Molecular Weight: 136,13 g/mol
Siedepunkt: 272 °C (1013 hPa)
Schmelzpunkt: 127 °C
MDL: MFCD00015477
CAS-Nummer: 77326-36-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR