2-Amino-3,5-dibrombenzonitril

Lieferant: CHEMPUR

SEW04283.250 SEW04283.1
CHMPSEW04283.250EA 0 EUR
CHMPSEW04283.250 CHMPSEW04283.1
2-Amino-3,5-dibrombenzonitril
2-Amino-3,5-dibrombenzonitril
Formel: C₇H₄Br₂N₂
Molecular Weight: 275,93 g/mol
MDL: MFCD00179571
CAS-Nummer: 68385-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR