2-Cyan-4,6-dichloranilin

Lieferant: CHEMPUR

SEW04284.1 SEW04284.10
CHMPSEW04284.1EA 0 EUR
CHMPSEW04284.1 CHMPSEW04284.10
2-Cyan-4,6-dichloranilin
2-Cyan-4,6-dichloranilin
Formel: C₇H₄Cl₂N₂
Molecular Weight: 187.03 g/mol
MDL: MFCD00158947
CAS-Nummer: 36764-94-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR