2-Cyan-4-fluoranilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

642924-10G 642924-1G
SIAL642924-10GEA 248 EUR
SIAL642924-10G SIAL642924-1G
2-Cyan-4-fluoranilin, Sigma-Aldrich®
2-Cyan-4-fluoranilin
Formel: C₇H₅FN₂
Molecular Weight: 136.13 g/mol
MDL: MFCD02683869
CAS-Nummer: 61272-77-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR