2-Cyan-4-nitroanilin 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Nitroanthranilonitril, 2-Amino-5-nitrobenzoesäurenitril, 2-Amino-5-nitrobenzonitril

B25416.14 B25416.22 B25416.36
B25416.14EA 33.8 EUR
B25416.14 B25416.22 B25416.36
2-Cyan-4-nitroanilin 95%
2-Cyan-4-nitroanilin

Purity: 95% CAS No.: 17420-30-3 Molecular Formula: C7H5N3O2 Formula Weight: 163.14 Melting Point: 207-210° MDL No.: MFCD00007362

Formel: C₇H₅N₃O₂
Schmelzpunkt: 207…210 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007362
CAS-Nummer: 17420-30-3
EINECS: 241-446-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR