2-Cyan-5-methylanilin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Cyano-5-methylaniline
43260.03
43260.14
43260.03EA
51.5
EUR
43260.03
43260.14
2-Cyan-5-methylanilin ≥98%
2-Cyan-5-methylanilin
Purity: 98%
CAS No.: 26830-96-6
Molecular Formula: C8H8N2
Formula Weight: 132.17
Physical Form: Powder
Melting Point: 91-93°
UN No.: 3439
MDL No.: MFCD00173706
Formel:
C₈H₈N₂ Molecular Weight: 132,16 g/mol Schmelzpunkt: 91…93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173706 CAS-Nummer: 26830-96-6 EINECS: 248-020-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...