2-Cyan-5-methylanilin

Lieferant: Thermo Scientific

Synonyme: 2-Cyano-5-methylaniline

TL00552DA TL00552EA
MAYBTL00552DAEA 0 EUR
MAYBTL00552DA MAYBTL00552EA
2-Cyan-5-methylanilin
2-Cyan-5-methylanilin
Formel: C₈H₈N₂
Molecular Weight: 132,16 g/mol
MDL: MFCD00173706
CAS-Nummer: 26830-96-6
EINECS: 248-020-0
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR