3-Cyan-4-fluoranilin 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 5-Amino-2-fluorbenzonitril, 5-Amino-2-fluorbenzoesäurenitril

H26210.06 H26210.14
H26210.06EA 39.7 EUR
H26210.06 H26210.14
3-Cyan-4-fluoranilin 97%
3-Cyan-4-fluoranilin

Purity: 97% CAS No.: 53312-81-5 Molecular Formula: C7H5FN2 Formula Weight: 136.13 Melting Point: 92-96° UN No.: 3439 MDL No.: MFCD00277872

Formel: C₇H₅FN₂
Molecular Weight: 136.13 g/mol
Schmelzpunkt: 92…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00277872
CAS-Nummer: 53312-81-5
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR