3-Cyan-4-fluoranilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Amino-2-fluorbenzonitril, 5-Amino-2-fluorbenzoesäurenitril

M-Clarity™ Qualitätsstufe = MQ100

639877-25G 639877-5G
SIAL639877-25GEA 264 EUR
SIAL639877-25G SIAL639877-5G
3-Cyan-4-fluoranilin, Sigma-Aldrich®
3-Cyan-4-fluoranilin
Formel: C₇H₅FN₂
Molecular Weight: 136,13 g/mol
MDL: MFCD00277872
CAS-Nummer: 53312-81-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR