3-Cyan-4-fluoranilin
Lieferant: FLUOROCHEM
Synonyme:
5-Amino-2-fluorbenzonitril, 5-Amino-2-fluorbenzoesäurenitril
013105-5G
013105-25G
013105-100G
FLUO013105-5GEA
31.1
EUR
FLUO013105-5G
FLUO013105-25G
FLUO013105-100G
3-Cyan-4-fluoranilin
3-Cyan-4-fluoranilin
Formel:
C₇H₅FN₂ Molecular Weight: 136.13 g/mol |
MDL:
MFCD00277872 CAS-Nummer: 53312-81-5 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...