1-Butylpyridiniumbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-butylpyridinium bromide, 1-n-Butylpyridinium bromide

M-Clarity™ Qualitätsstufe = MQ100

00285-50G-F 00285-5G-F
SIAL00285-50G-FEA 211 EUR
SIAL00285-50G-F SIAL00285-5G-F
1-Butylpyridiniumbromid, Sigma-Aldrich®
1-Butylpyridinium bromide
Formel: C₉H₁₄BrN
Molecular Weight: 216,12 g/mol
MDL: MFCD00671496
CAS-Nummer: 874-80-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR