1-Butylpyridiniumchlorid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-Butylpyridinium Chloride, N-Butylpyridinium chloride

19740.04 19740.09 19740.18
19740.04EA 52.6 EUR
19740.04 19740.09 19740.18
1-Butylpyridiniumchlorid ≥98%
1-Butylpyridinium chloride

Purity: 98% CAS No.: 1124-64-7 Molecular Formula: C9H14ClN Formula Weight: 171.67 Physical Form: Crystalline Sensitivity: Hygroscopic MDL No.: MFCD00060047

Formel: C₉H₁₄ClN
Molecular Weight: 171.67 g/mol
Schmelzpunkt: 131…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00060047
CAS-Nummer: 1124-64-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR