2-Fluor-4-nitrobenzonitril

Lieferant: CHEMPUR

FL13092.POR 013092-25G 013092-5G 013092-10G 013092-20G
CHMPFL13092.POREA 0 EUR
CHMPFL13092.POR FLUO013092-25G FLUO013092-5G FLUO013092-10G FLUO013092-20G
2-Fluor-4-nitrobenzonitril
2-Fluor-4-nitrobenzonitril
Formel: C₇H₃FN₂O₂
Molecular Weight: 166,11 g/mol
MDL: MFCD04038715
CAS-Nummer: 34667-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR