2-Fluor-5-nitrobenzonitril ≥98%

Lieferant: Thermo Scientific
Gefahr

A12417.03 A12417.06
A12417.03EA 61.3 EUR
A12417.03 A12417.06
2-Fluor-5-nitrobenzonitril ≥98%
2-Fluor-5-nitrobenzonitril

Purity: 98+% CAS No.: 17417-09-3 Molecular Formula: C7H3FN2O2 Formula Weight: 166.11 Melting Point: 70-72° Beilstein Reference: 2048720 UN No.: 3439 MDL No.: MFCD00042299

Formel: C₇H₃FN₂O₂
Molecular Weight: 166,11 g/mol
Schmelzpunkt: 70…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042299
CAS-Nummer: 17417-09-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR