2-Fluor-5-nitrobenzonitril

Lieferant: Thermo Scientific

TL00351DA TL00351EA
MAYBTL00351DAEA 0 EUR
MAYBTL00351DA MAYBTL00351EA
2-Fluor-5-nitrobenzonitril
2-Fluor-5-nitrobenzonitril
Formel: C₇H₃FN₂O₂
Molecular Weight: 166,11 g/mol
Schmelzpunkt: 79 °C
MDL: MFCD00042299
CAS-Nummer: 17417-09-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR