2-Fluor-6-hydroxybenzonitril

Lieferant: FLUOROCHEM

Synonyme: 2-Fluor-6-hydroxybenzoesäurenitril

013036-1G 013036-5G
FLUO013036-1GEA 36.9 EUR
FLUO013036-1G FLUO013036-5G
2-Fluor-6-hydroxybenzonitril
2-Fluor-6-hydroxybenzonitril
Formel: C₇H₄FNO
Molecular Weight: 137.11 g/mol
MDL: MFCD03428592
CAS-Nummer: 140675-43-0
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR