2-Fluor-6-iodbenzonitril

Lieferant: FLUOROCHEM

006244-1G 006244-5G 006244-25G
FLUO006244-1GEA 31.1 EUR
FLUO006244-1G FLUO006244-5G FLUO006244-25G
2-Fluor-6-iodbenzonitril
2-Fluor-6-iodbenzonitril
Formel: C₇H₃FIN
Molecular Weight: 247.01 g/mol
Siedepunkt: 281
Schmelzpunkt: 48.5…50.5 °C
MDL: MFCD00015478
CAS-Nummer: 79544-29-9
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR