4-Nitro-o-toluonitril, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Methyl-4-nitrobenzonitril, 2-Methyl-4-nitrobenzoesäurenitril
M-Clarity™ Qualitätsstufe = MQ100
537012-1G
537012-5G
SIAL537012-1GEA
36
EUR
SIAL537012-1G
SIAL537012-5G
4-Nitro-o-toluonitril, Sigma-Aldrich®
4-Nitro-o-toluonitril
Formel:
C₈H₆N₂O₂ Molecular Weight: 162,15 g/mol |
MDL:
MFCD03095378 CAS-Nummer: 89001-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...