5-Nitro-o-toluonitril
Lieferant: Thermo Scientific
Synonyme:
2-Methyl-5-nitrobenzoesäurenitril, 2-Methyl-5-nitrobenzonitril
BTB09853.1
BTB09853.10
BTB09853.50
MAYBBTB09853.1EA
0
EUR
MAYBBTB09853.1
MAYBBTB09853.10
MAYBBTB09853.50
5-Nitro-o-toluonitril
5-Nitro-o-toluonitril
Formel:
C₈H₆N₂O₂ Molecular Weight: 162,15 g/mol Siedepunkt: 174 °C (18 mmHg) Schmelzpunkt: 104…106 °C |
MDL:
MFCD00017040 CAS-Nummer: 939-83-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...