6-Nitro-o-toluonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methyl-6-nitrobenzoesäurenitril, 2-Methyl-6-nitrobenzonitril

M-Clarity™ Qualitätsstufe = MQ100

188719-25G 188719-5G
SIAL188719-25GEA 387 EUR
SIAL188719-25G SIAL188719-5G
6-Nitro-o-toluonitril, Sigma-Aldrich®
6-Nitro-o-toluonitril
Formel: C₈H₆N₂O₂
Molecular Weight: 162,15 g/mol
MDL: MFCD00007266
CAS-Nummer: 1885-76-3
EINECS: 217-553-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR