DL-α-Phenylalanin ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
(±)-2-Amino-3-phenylpropansäure, H-DL-Phe-OH, 3-Phenyl-DL-alanin, DL-Phenylalanin, (±)-2-Amino-3-phenylpropionic acid, (±)-2-Amino-3-phenylpropionsäure
A10132.14
A10132.22
A10132.36
A10132.14EA
28.5
EUR
A10132.14
A10132.22
A10132.36
DL-α-Phenylalanin ≥99%
DL-α-Phenylalanin
Purity: 99%
CAS No.: 150-30-1
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Melting Point: ca 266° dec.
Beilstein Reference: 1910407
Merck Reference: 14,7271
MDL No.: MFCD00064225
Formel:
C₉H₁₁NO₂ Molecular Weight: 165,19 g/mol Siedepunkt: 313 °C (1013 hPa) Schmelzpunkt: ∼ 266 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064225 CAS-Nummer: 150-30-1 EINECS: 205-756-7 Merck Index: 14,7271 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...