Cole-Parmer® Essentials Silicone Stoppers
Lieferant: Cole-Parmer
06298-06 06298-22 06298-18
COLE06298-06EA 0 EUR
COLE06298-06 COLE06298-22 COLE06298-18
Cole-Parmer® Essentials Silicone Stoppers
Stopfen Silikonstopfen
Silicone bubbles make these stoppers lightweight-they float on water.
  • Resistant to temperatures from −60 to +200 °C
Softness and flexibility allow stoppers to fit a range of holes.