2-Chlor-5-(chlormethyl)pyridin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
516910-25G
516910-5G
SIAL516910-25GEA
70.8
EUR
SIAL516910-25G
SIAL516910-5G
2-Chlor-5-(chlormethyl)pyridin, Sigma-Aldrich®
2-Chloro-5-(chloromethyl)pyridine
Formel:
C₆H₅Cl₂N Molecular Weight: 162.02 g/mol Schmelzpunkt: 37 |
MDL:
MFCD00125366 CAS-Nummer: 70258-18-3 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...