2-Chlor-5-(chlormethyl)pyridin
Lieferant: FLUOROCHEM
019460-10G
019460-50G
FLUO019460-10GEA
0
EUR
FLUO019460-10G
FLUO019460-50G
2-Chlor-5-(chlormethyl)pyridin
2-Chloro-5-(chloromethyl)pyridine
Formel:
C₆H₅Cl₂N Schmelzpunkt: 37 °C |
MDL:
MFCD00125366 CAS-Nummer: 70258-18-3 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...