2-Fluorisobuttersäure 95%

Lieferant: Thermo Scientific
Gefahr

H25837.MD H25837.03
H25837.MDEA 50.7 EUR
H25837.MD H25837.03
2-Fluorisobuttersäure 95%
2-Fluorisobuttersäure

Purity: 95% CAS No.: 63812-15-7 Molecular Formula: C4H7FO2 Formula Weight: 106.10 Refractive Index: 1.3850 Boiling Point: 57-58°/10mm UN No.: 2922 MDL No.: MFCD03452788

Formel: C₄H₇FO₂
Molecular Weight: 106,1 g/mol
Siedepunkt: 57…58 °C (10 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD03452788
CAS-Nummer: 63812-15-7
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR