2-Fluorisobuttersäure

Lieferant: FLUOROCHEM

013150-1G 013150-5G
FLUO013150-1GEA 45.9 EUR
FLUO013150-1G FLUO013150-5G
2-Fluorisobuttersäure
2-Fluorisobuttersäure
Formel: C₄H₇FO₂
Molecular Weight: 106,1 g/mol
MDL: MFCD03452788
CAS-Nummer: 63812-15-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR