5-Brom-2,3-diaminopyridin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Bromopyridine-2,3-diyldiamine, 2,3-Diamino-5-bromopyridine

350330050 350330250
ACRO350330050EA 16.4 EUR
ACRO350330050 ACRO350330250
5-Brom-2,3-diaminopyridin 97%
5-Brom-2,3-diaminopyridin
Formel: C₅H₆BrN₃
Molecular Weight: 188,03 g/mol
Siedepunkt: 321 °C (1013 hPa)
Schmelzpunkt: 160…165 °C
MDL: MFCD00460094
CAS-Nummer: 38875-53-5
EINECS: 254-172-9
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to light brown to purple Powder or needles
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR