5-Brom-2,3-diaminopyridin 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 5-Bromopyridine-2,3-diyldiamine, 2,3-Diamino-5-bromopyridine

L19435.06 L19435.14
L19435.06EA 16 EUR
L19435.06 L19435.14
5-Brom-2,3-diaminopyridin 97%
5-Brom-2,3-diaminopyridin

Purity: 97% CAS No.: 38875-53-5 Molecular Formula: C5H6BrN3 Formula Weight: 188.03 Melting Point: 153-161° Beilstein Reference: 119436 UN No.: 2811 MDL No.: MFCD00460094

Formel: C₅H₆BrN₃
Molecular Weight: 188.03 g/mol
Siedepunkt: 321
Schmelzpunkt: 153…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00460094
CAS-Nummer: 38875-53-5
EINECS: 254-172-9
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR