(±)-3-Hydroxybuttersäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(±)-3-Hydroxybutansäure
H30426.06
H30426.14
H30426.06EA
90.1
EUR
H30426.06
H30426.14
(±)-3-Hydroxybuttersäure ≥98%
(±)-3-Hydroxybuttersäure
Purity: 98%
CAS No.: 300-85-6
Molecular Formula: C4H8O3
Formula Weight: 104.10
Odour: Odourless
Melting Point: 36-39°
MDL No.: MFCD00004546
Formel:
C₄H₈O₃ Molecular Weight: 104,11 g/mol Siedepunkt: 269 °C (1013 hPa) Schmelzpunkt: 36…39 °C Dichte: 1,26 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004546 CAS-Nummer: 300-85-6 EINECS: 206-099-9 Merck Index: 14,4816 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...