3-Pyridinboronsäure

Lieferant: CHEMPUR

Synonyme: 3-Pyridinylboronsäure, Pyridin-3-boronsäure

CC04112.0.1 CC04112.250
CHMPCC04112.0.1EA 0 EUR
CHMPCC04112.0.1 MAYBCC04112.250
3-Pyridinboronsäure
3-Pyridineboronic acid
Formel: C₅H₆BNO₂
Molecular Weight: 122.92 g/mol
MDL: MFCD00674177
CAS-Nummer: 1692-25-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR