4-Pyridinboronsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Pyridinylboronsäure, Pyridin-4-boronsäure
M-Clarity™ Qualitätsstufe = MQ100
634492-1G
634492-25G
634492-5G
SIAL634492-1GEA
37.6
EUR
SIAL634492-1G
SIAL634492-25G
SIAL634492-5G
4-Pyridinboronsäure, Sigma-Aldrich®
4-Pyridineboronic acid
Formel:
C₅H₆BNO₂ Molecular Weight: 122,92 g/mol (anhy) |
MDL:
MFCD01074545 CAS-Nummer: 1692-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...