2-Fluor-5-pyridinboronsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Fluorpyridin-5-boronsäure, 2-Fluor-5-pyridinylboronsäure
M-Clarity™ Qualitätsstufe = MQ100
639184-1G
639184-5G
SIAL639184-1GEA
22.8
EUR
SIAL639184-1G
SIAL639184-5G
2-Fluor-5-pyridinboronsäure, Sigma-Aldrich®
2-Fluor-5-pyridinboronsäure
Formel:
C₅H₅BFNO₂ Molecular Weight: 140,91 g/mol |
MDL:
MFCD03411559 CAS-Nummer: 351019-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...