Heptafluorbuttersäureanhydrid Acylierungsreagenz, zur GC-Derivatisierung
Lieferant: MACHEREY-NAGEL
Gefahr
Synonyme: Perfluorbuttersäureanhydrid, HFBA, HFAA
701110.201 701110.110 701110.510
MANA701110.201EA 275 EUR
MANA701110.201 MANA701110.110 MANA701110.510
Heptafluorbuttersäureanhydrid Acylierungsreagenz, zur GC-Derivatisierung
Heptafluorbuttersäureanhydrid
Acylation with HFBA can be used for alcohols, phenols, carboxylic acids, amines, amino acids and steroids forming volatile, stable derivatives suited for GC-FID as well as for GC-ECD detection.
  • Addition of halogenated carbons enhances detectability by GC-ECD
  • Derivatives are hydrolytical stable
  • Increased sensitivity by adding molecular weight
Formel: C₈F₁₄O₃
Molecular Weight: 410,06 g/mol
Siedepunkt: 108 °C (1013 hPa)
Schmelzpunkt: 14 °C
Dichte: 1,653 g/cm³ (25 °C)
MDL: MFCD00000432
CAS-Nummer: 336-59-4
EINECS: 206-410-8
UN: 3316
ADR: 9,II