Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate 98%, rein
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Ethylenediaminetetraaceticacidferricsodiumsalttrihydrate, (Ethylenedinitrilo)tetraacetic acid ferric sodium salt, EDTA ferric sodium salt
304680010
304682500
ACRO304680010EA
359
EUR
ACRO304680010
ACRO304682500
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate 98%, rein
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate
Formel:
C₁₀H₁₂FeN₂NaO₈·3H₂O Molecular Weight: 421,09 g/mol Schmelzpunkt: 80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798117 CAS-Nummer: 18154-32-0 EINECS: 239-802-2 Merck Index: 12,04076 |
Spezifikation Testergebnisse
Appearance | Yellow-brown Powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...