2,4-Difluor-N-methylanilin

Lieferant: FLUOROCHEM

019202-10G 019202-25G
FLUO019202-10GEA 0 EUR
FLUO019202-10G FLUO019202-25G
2,4-Difluor-N-methylanilin
2,4-Difluor-N-methylanilin
Formel: C₇H₇F₂N
Molecular Weight: 143.14 g/mol
Siedepunkt: 94…96 °C (15 mmHg)
MDL: MFCD00042472
CAS-Nummer: 138564-16-6
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR