2-Brom-4,6-di-tert-butylphenol

Lieferant: Thermo Scientific

BTB04137.1 BTB04137.10
MAYBBTB04137.1EA 0 EUR
MAYBBTB04137.1 MAYBBTB04137.10
2-Brom-4,6-di-tert-butylphenol
2-Brom-4,6-di-tert-butylphenol
Formel: C₁₄H₂₁BrO
Molecular Weight: 285,22 g/mol
MDL: MFCD00051597
CAS-Nummer: 20834-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR