4-Brom-2,6-di-tert-butylphenol
Lieferant: Thermo Scientific
Synonyme:
1-Bromo-3,5-di(dimethylethyl)-4-hydroxybenzene
BTB06414.1
BTB06414.10
BTB06414.50
MAYBBTB06414.1EA
0
EUR
MAYBBTB06414.1
MAYBBTB06414.10
MAYBBTB06414.50
4-Brom-2,6-di-tert-butylphenol
4-Brom-2,6-di-tert-butylphenol
Formel:
C₁₄H₂₁BrO Molecular Weight: 285,22 g/mol Siedepunkt: 104…107 °C (1 mmHg) Schmelzpunkt: 83 °C |
MDL:
MFCD00051598 CAS-Nummer: 1139-52-2 EINECS: 214-521-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...