4-Methyl-2,3,5,6-tetrabromphenol

Lieferant: FLUOROCHEM

Synonyme: 4-Hydroxy-2,3,5,6-tetrabromtoluol, 2,3,5,6-Tetrabrom-p-kresol

018992-1G 018992-5G
FLUO018992-1GEA 45.9 EUR
FLUO018992-1G FLUO018992-5G
4-Methyl-2,3,5,6-tetrabromphenol
4-Methyl-2,3,5,6-tetrabromphenol
Formel: C₇H₄Br₄O
Molecular Weight: 423.72 g/mol
Siedepunkt: 356
Schmelzpunkt: 209
MDL: MFCD00060318
CAS-Nummer: 37721-75-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR