Methylrot Natriumsalz ACS

Lieferant: Thermo Scientific

Synonyme: 2-(4-(Dimethylamino)phenylazo)benzoesäure Natriumsalz, Methylrot wasserlöslich, Natrium-2-(p-(dimethylamino)phenylazo)benzoat

414840100 414840250 414841000
ACRO414840100EA 18.4 EUR
ACRO414840100 ACRO414840250 ACRO414841000
Methylrot Natriumsalz ACS
Methylrot Natriumsalz
Formel: C₁₅H₁₄N₃NaO₂
Molecular Weight: 291,28 g/mol
Schmelzpunkt: > 180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002426
CAS-Nummer: 845-10-3
EINECS: 212-682-9
Merck Index: 13,06142

Bestellen

Spezifikation Testergebnisse

Appearance Red to brown Powder
Infrared spectrum Conforms
Visual transition interval From pH 4.2 (pink) to pH 6.2 (yellow)
Clarity of solution (Aqueous) Passes test
Clarity of alcohol solution Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR