Methylrot 1 g/l in Ethanol

Lieferant: Bernd Kraft
Gefahr

Synonyme: 2-(4-(Dimethylamino)phenylazo)benzoesäure

03091.1700 03091.3000
KRAF03091.1700EA 35.8 EUR
KRAF03091.1700 KRAF03091.3000
Methylrot 1 g/l in Ethanol
Methylrot
Formel: C₁₅H₁₅N₃O₂
Molecular Weight: 269.3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002425
CAS-Nummer: 493-52-7
EINECS: 207-776-1
UN: 1170
ADR: 3 II
Merck Index: 12,06199

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR