2-Acetyl-5-methylthiophen ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-(5-Methyl-2-thienyl)-1-ethanone

A11282.06 A11282.14 A11282.22
A11282.06EA 61.9 EUR
A11282.06 A11282.14 A11282.22
2-Acetyl-5-methylthiophen ≥98%
2-Acetyl-5-methylthiophen

Purity: 98% CAS No.: 13679-74-8 Molecular Formula: C7H8OS Formula Weight: 140.20 Storage Temperature: 2°C to 8°C Refractive Index: 1.5625 Melting Point: 24-28° Boiling Point: 65-67°/1mm Beilstein Reference: 110854 MDL No.: MFCD00014529

Formel: C₇H₈OS
Molecular Weight: 140,21 g/mol
Siedepunkt: 65…67 °C (1 mmHg)
Schmelzpunkt: 24…28 °C
Dichte: 1,119
MDL: MFCD00014529
CAS-Nummer: 13679-74-8
EINECS: 237-181-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR