2-Thiophenboronsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Thienylboronsäure, Thiophen-2-boronsäure
344710010
344710050
344710250
ACRO344710010EA
33.3
EUR
ACRO344710010
ACRO344710050
ACRO344710250
2-Thiophenboronsäure 97%
2-Thiopheneboronic acid
Formel:
C₄H₅BO₂S Molecular Weight: 127,96 g/mol Schmelzpunkt: 120…122 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00151850 CAS-Nummer: 6165-68-0 |
Spezifikation Testergebnisse
Appearance | Off-white to beige Powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...