2-Thiophenboronsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Thienylboronsäure, Thiophen-2-boronsäure

B23071.06 B23071.03 B23071.14
B23071.06EA 109 EUR
B23071.06 B23071.03 B23071.14
2-Thiophenboronsäure ≥98%
2-Thiopheneboronic acid

Purity: 98% CAS No.: 6165-68-0 Molecular Formula: C4H5BO2S Formula Weight: 127.96 Storage Temperature: 2°C to 8°C Melting Point: 128-131° Beilstein Reference: 112375 MDL No.: MFCD00151850

Formel: C₄H₅BO₂S
Molecular Weight: 127,96 g/mol
Schmelzpunkt: 128…131 °C
MDL: MFCD00151850
CAS-Nummer: 6165-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR