Bromphenolblau Natriumsalz ACS

Lieferant: Thermo Fisher Scientific

Synonyme: 4,4'-(3H-2,1-Benzoxathiol-3-yliden)bis[2,6-dibromphenol]-S,S-dioxid Natriumsalz, Bromphenolblau wasserlöslich, 3,3',5,5'-Tetrabromphenolsulfonphthalein Natriumsalz, Natrium-α-(3,5-dibrom-4-oxo-2,5-cyclohexadienyliden)-α-(3,5-dibrom-4-hydroxyphenyl)toluolsulfonat

32639.06 32639.14 32639.22
32639.06EA 35.5 EUR
32639.06 32639.14 32639.22
Bromphenolblau Natriumsalz ACS
Bromphenolblau Natriumsalz

CAS No.: 34725-61-6 Molecular Formula: C19H9Br4NaO5S Formula Weight: 691.96 Physical Form: Crystalline Melting Point: 300° Merck Reference: 14,1444 MDL No.: MFCD00013793

Formel: C₁₉H₉Br₄NaO₅S
Molecular Weight: 691.95 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013793
CAS-Nummer: 34725-61-6
EINECS: 252-170-2
Merck Index: 14,1444

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR