Bromphenolblau Natriumsalz
Lieferant: Serva
Synonyme:
4,4'-(3H-2,1-Benzoxathiol-3-yliden)bis[2,6-dibromphenol]-S,S-dioxid Natriumsalz, Bromphenolblau wasserlöslich, 3,3',5,5'-Tetrabromphenolsulfonphthalein Natriumsalz, Natrium-α-(3,5-dibrom-4-oxo-2,5-cyclohexadienyliden)-α-(3,5-dibrom-4-hydroxyphenyl)toluolsulfonat
15375.01
15375.02
SERA15375.01EA
39.7
EUR
SERA15375.01
SERA15375.02
Bromphenolblau Natriumsalz
Bromphenolblau Natriumsalz
Formel:
C₁₉H₉Br₄NaO₅S Molecular Weight: 691,95 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013793 CAS-Nummer: 34725-61-6 EINECS: 252-170-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...