Bromphenolblau Natriumsalz für die Molekularbiologie, geeignet für die Elektrophorese, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 4,4'-(3H-2,1-Benzoxathiol-3-yliden)bis[2,6-dibromphenol]-S,S-dioxid Natriumsalz, Bromphenolblau wasserlöslich, 3,3',5,5'-Tetrabromphenolsulfonphthalein Natriumsalz, Natrium-α-(3,5-dibrom-4-oxo-2,5-cyclohexadienyliden)-α-(3,5-dibrom-4-hydroxyphenyl)toluolsulfonat

M-Clarity™ Qualitätsstufe = MQ200

B8026-25G B8026-5G
B8026-25GEA 285 EUR
B8026-25G B8026-5G
Bromphenolblau Natriumsalz für die Molekularbiologie, geeignet für die Elektrophorese, Sigma-Aldrich®
Bromphenolblau Natriumsalz

Bromophenol blue (BroB), an arylmethane color marker dye has a molecular mass of 670 Da.


  • Bromophenol Blue sodium salt has been used in the buffer solution to differentiate dead and living cell
  • It has also been used as a constituent in gel loading buffer
  • A tracking dye for nucleic acid or protein electrophoresis in agarose or polyacrylamide gels

Formel: C₁₉H₉Br₄NaO₅S
Lagertemperatur: Room Temperature
MDL: MFCD00013793
CAS-Nummer: 34725-61-6
EINECS: 252-170-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce5c8fd34a097127aac7cdacb27e7f410130de5e&vid=ec79008533fdd3afb43a601833c6d55c02011407&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form) Powder
Suitability
Suitable For Use as a Tracking Dye
For Electrophoresis in Alkaline and Neutral Buffers
Suitable
Visual Interval
pH 3.0 (Yellow) - pH 4.6 (Blue)
Pass
Clarity of Solution Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR