Bromthymolblau 0,1% in Ethanol

Lieferant: Bernd Kraft
Gefahr

Synonyme: 3′,3′′-Dibromthymolsulfonphthalein, BTB

03118.3000 03118.2000
KRAF03118.3000EA 87.7 EUR
KRAF03118.3000 KRAF03118.2000
Bromthymolblau 0,1% in Ethanol
Bromthymolblau
Formel: C₂₇H₂₈Br₂O₅S
Molecular Weight: 624.39 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005872
CAS-Nummer: 76-59-5
EINECS: 200-971-2
UN: 1170
ADR: 3 II
Merck Index: 13,01430

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR